Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182973.1 | NP_892018.1 | 12 | 1973 | ||
XM_011527978.2 | XP_011526280.1 | 14 | 2336 | |||
XM_011527980.1 | XP_011526282.1 | 6 | 992 | |||
XM_011527982.2 | XP_011526284.1 | 4 | 767 | |||
GenBank mRNA | AJ488946.1 | - | 12 | 1973 | ||
AJ488947.1 | - | 12 | 1973 |