Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290307.2 | NP_001277236.1 | 2, 3 | 240 | ||
NM_001290310.2 | NP_001277239.1 | 2, 3 | 240 | |||
NM_001323982.1 | NP_001310911.1 | 3, 4 | 710 | |||
NM_001323983.1 | NP_001310912.1 | 2, 3 | 392 | |||
NM_001323984.1 | NP_001310913.1 | 3, 4 | 258 | |||
NM_001323985.1 | NP_001310914.1 | 2, 3 | 240 | |||
NM_001323986.1 | NP_001310915.1 | 2, 3 | 240 | |||
NM_001903.4 | NP_001894.2 | 2, 3 | 240 | |||
GenBank mRNA | AK094151.1 | - | 2, 3 | 179 | ||
AK296708.1 | - | 2, 3 | 211 | |||
AK304267.1 | - | 2, 3 | 180 | |||
BC000385.2 | - | 2, 3 | 178 | 2821111 | ||
D13866.1 | - | 2, 3 | 179 | |||
D14705.1 | - | 2, 3 | 204 | |||
DC332608.1 | - | 2, 3 | 240 | |||
DC369291.1 | - | 2, 3 | 230 | |||
L23805.1 | - | 1, 2 | 117 | |||
U03100.1 | - | 1, 2 | 117 |