Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008709.1 | NP_001008709.1 | 5 | 878 | ||
NM_002708.3 | NP_002699.1 | 5 | 845 | |||
NM_206873.1 | NP_996756.1 | 4 | 713 | |||
GenBank mRNA | AK098311.1 | - | 7 | 1654 | ||
AK127616.1 | - | 6 | 1062 | |||
AK313586.1 | - | 5 | 754 | |||
AL557336.3 | - | 5 | 869 | |||
BC001888.1 | - | 5 | 766 | 3546588 | ||
BC004482.2 | - | 5 | 786 | 3689198 | ||
BC008010.1 | - | 5 | 786 | 3528742 | ||
BM799599.1 | - | 4 | 652 | |||
BT006629.1 | - | 5 | 722 | |||
J04759.1 | - | 4 | 656 | |||
M63960.1 | - | 5 | 751 | |||
S57501.1 | - | 5 | 766 | |||
X70848.1 | - | 5 | 749 |