Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP2R5C SIRNA Assays ›
Gene Symbol:
PPP2R5C
Gene Name:
protein phosphatase 2 regulatory subunit B'gamma
Gene Aliases:
B56G, PR61G
Chromosome Location:
Chr.14: 101760573 - 101927992 on Build GRCh38
UniGene ID:
Hs.368264
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
105832
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP2R5C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001161725.1
NP_001155197.1
4
332
NM_001161726.1
NP_001155198.1
4
404
NM_002719.3
NP_002710.2
2
283
NM_178586.2
NP_848701.1
2
283
NM_178587.2
NP_848702.1
2
283
XM_005267819.1
XP_005267876.1
4
393
XM_005267820.1
XP_005267877.1
4
333
XM_005267822.3
XP_005267879.1
4
393
XM_005267823.1
XP_005267880.1
4
393
XM_005267824.1
XP_005267881.1
4
393
XM_005267826.1
XP_005267883.1
2
106
XM_005267827.1
XP_005267884.1
4
328
XM_011536914.2
XP_011535216.1
3
197
XM_011536915.2
XP_011535217.1
3
280
XM_011536916.2
XP_011535218.1
3
1279
XM_011536918.1
XP_011535220.1
3
434
XM_011536919.2
XP_011535221.1
3
416
XM_011536920.1
XP_011535222.1
3
310
XM_011536921.2
XP_011535223.1
2
199
XM_017021422.1
XP_016876911.1
3
349
XM_017021423.1
XP_016876912.1
2
283
XM_017021424.1
XP_016876913.1
3
310
GenBank mRNA
AB451341.1
-
2
143
AI692258.1
-
4
405
AK131391.1
-
4
317
AK302470.1
-
4
232
AK315072.1
-
2
219
BC016183.1
-
2
181
4855950
BI560029.1
-
2
257
D26445.1
-
2
140
DA853338.1
-
2
283
L42375.1
-
2
113
U37352.1
-
2
201
Z69030.1
-
2
221
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00604902_m1
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
negative regulation of cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
regulation of protein phosphatase type 2A activity
proteasome-mediated ubiquitin-dependent protein catabolic process
protein binding
protein phosphatase type 2A regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-77psw:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline