Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006741.3 | NP_006732.3 | 2, 3 | 326 | ||
XM_005268995.4 | XP_005269052.1 | 2, 3 | 306 | |||
XM_006719471.3 | XP_006719534.1 | 2, 3 | 320 | |||
GenBank mRNA | AK123969.1 | - | 3, 4 | 251 | ||
AY063766.1 | - | 2, 3 | 157 | |||
BC022470.1 | - | 2, 3 | 326 | 4819907 | ||
BP211831.1 | - | 2, 3 | 326 | |||
CR457003.1 | - | 2, 3 | 155 | |||
U48707.1 | - | 2, 3 | 163 |