Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014214.2 | NP_055029.1 | 2 | 463 | ||
XM_011525661.2 | XP_011523963.1 | 2 | 426 | |||
GenBank mRNA | AF014398.2 | - | 2 | 463 | ||
AF200432.1 | - | 1 | 140 | |||
BC017176.1 | - | 2 | 303 | 3345789 | ||
BC031560.1 | - | 2 | 375 | |||
BT007061.1 | - | 2 | 221 | |||
CF134610.1 | - | 2 | 420 |