Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319658.1 | NP_001306587.1 | 2 | 538 | ||
NM_001319659.1 | NP_001306588.1 | 3 | 497 | |||
NM_017823.4 | NP_060293.2 | 3 | 490 | |||
GenBank mRNA | AB164404.1 | - | 2 | 523 | ||
AF086553.1 | - | 2 | 338 | |||
AK000449.1 | - | 3 | 474 | |||
BC001140.1 | - | 3 | 460 | 3535406 | ||
BG823348.1 | - | 2 | 520 | |||
BQ679163.1 | - | 3 | 509 | |||
DA428735.1 | - | 2 | 541 |