Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286555.1 | NP_001273484.1 | 3 | 555 | ||
NM_020185.4 | NP_064570.1 | 3 | 555 | |||
NR_104473.1 | 3 | 555 | ||||
XM_017011061.1 | XP_016866550.1 | 3 | 153 | |||
GenBank mRNA | AF165519.1 | - | 3 | 96 | ||
AF424702.1 | - | 3 | 155 | |||
AK296402.1 | - | 3 | 125 | |||
AK310653.1 | - | 3 | 135 | |||
AY249859.1 | - | 3 | 559 | |||
BC022847.2 | - | 3 | 511 | 3942055 | ||
DA950312.1 | - | 3 | 138 |