Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286555.1 | NP_001273484.1 | 4 | 633 | ||
NM_020185.4 | NP_064570.1 | 4 | 633 | |||
NR_104474.1 | 3 | 550 | ||||
GenBank mRNA | AF165519.1 | - | 4 | 174 | ||
AF424702.1 | - | 4 | 233 | |||
AK296402.1 | - | 4 | 203 | |||
AY249859.1 | - | 4 | 637 | |||
BC022847.2 | - | 4 | 589 | 3942055 | ||
DB177174.1 | - | 3 | 87 |