Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167880.1 | NP_001161352.1 | 5 | 678 | ||
NM_001318332.1 | NP_001305261.1 | 5 | 678 | |||
NM_022126.3 | NP_071409.3 | 5 | 678 | |||
XM_005270026.3 | XP_005270083.1 | 5 | 627 | |||
XM_006717943.1 | XP_006718006.1 | 5 | 627 | |||
XM_011540058.2 | XP_011538360.1 | 5 | 627 | |||
XM_017016509.1 | XP_016871998.1 | 5 | 627 | |||
XM_017016510.1 | XP_016871999.1 | 5 | 627 | |||
XM_017016511.1 | XP_016872000.1 | 5 | 627 | |||
GenBank mRNA | AK055532.1 | - | 5 | 607 | ||
BC110344.1 | - | 5 | 632 | 5762731 | ||
BC113629.1 | - | 5 | 678 | 8322681 | ||
BC113631.1 | - | 5 | 678 | 8322683 | ||
BM541880.1 | - | 5 | 616 |