Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305115.1 | NP_001292044.1 | 2 | 625 | ||
NM_001305116.1 | NP_001292045.1 | 1 | 392 | |||
NM_024025.2 | NP_076930.1 | 2 | 628 | |||
GenBank mRNA | AB103376.1 | - | 2 | 443 | ||
AB158288.1 | - | 2 | 505 | |||
AB237597.1 | - | 2 | 440 | |||
AK055704.1 | - | 2 | 502 | |||
AY902194.1 | - | 2 | 443 | |||
BC001613.1 | - | 2 | 443 | 3535215 | ||
BC003115.1 | - | 1 | 14 | |||
BC067804.1 | - | 2 | 295 | 5262839 | ||
BI753746.1 | - | 1 | 324 | |||
BI834891.1 | - | 1 | 393 |