Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319210.1 | NP_001306139.1 | 3 | 647 | ||
NM_001319211.1 | NP_001306140.1 | 3 | 279 | |||
NM_001319212.1 | NP_001306141.1 | 3 | 647 | |||
NM_003672.3 | NP_003663.2 | 3 | 647 | |||
NM_033312.2 | NP_201569.1 | 3 | 647 | |||
NM_033313.2 | NP_201570.1 | 3 | 647 | |||
XM_005271294.3 | XP_005271351.1 | 3 | 187 | |||
XM_005271296.3 | XP_005271353.1 | 3 | 184 | |||
XM_011542340.2 | XP_011540642.1 | 3 | 186 | |||
XM_011542341.2 | XP_011540643.1 | 2 | 127 | |||
XM_011542346.2 | XP_011540648.1 | 3 | 195 | |||
XM_017002646.1 | XP_016858135.1 | 3 | 187 | |||
XM_017002647.1 | XP_016858136.1 | 3 | 728 | |||
GenBank mRNA | AF000367.1 | - | 3 | 514 | ||
AF064102.1 | - | 3 | 622 | |||
AF064103.1 | - | 3 | 612 | |||
AF122013.1 | - | 3 | 159 | |||
AK302310.1 | - | 3 | 268 | |||
AK310292.1 | - | 2 | 165 | |||
AK310657.1 | - | 3 | 605 | |||
BC038979.1 | - | 3 | 221 | |||
BC093916.1 | - | 3 | 254 | 7939761 | ||
BC093918.1 | - | 3 | 254 | 7939763 | ||
DA885382.1 | - | 3 | 476 | |||
DB029799.1 | - | 3 | 292 | |||
DQ530256.1 | - | 3 | 165 |