Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACYP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320586.1 | NP_001307515.1 | 7 | 694 | ||
NM_001320587.1 | NP_001307516.1 | 6 | 601 | |||
NM_001320588.1 | NP_001307517.1 | 4 | 395 | |||
NM_001320589.1 | NP_001307518.1 | 5 | 716 | |||
NM_138448.3 | NP_612457.1 | 4 | 646 | |||
XM_017005411.1 | XP_016860900.1 | 8 | 1040 | |||
XM_017005412.1 | XP_016860901.1 | 9 | 1110 | |||
XM_017005413.1 | XP_016860902.1 | 9 | 1081 | |||
GenBank mRNA | BC012290.1 | - | 4 | 415 | 3864319 | |
BF671963.1 | - | 4 | 394 | |||
BG741807.1 | - | 6 | 595 | |||
BU543943.1 | - | 7 | 694 | |||
BU857820.1 | - | 5 | 574 | |||
X84195.1 | - | 4 | 395 |