Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000073.2 | NP_000064.1 | 7 | 782 | ||
XM_006718941.2 | XP_006719004.1 | 7 | 765 | |||
GenBank mRNA | AK223314.1 | - | 7 | 735 | ||
AK310578.1 | - | 8 | 910 | |||
AK310622.1 | - | 8 | 940 | |||
BC113830.1 | - | 7 | 728 | 40029587 | ||
DB116263.1 | - | 4 | 349 | |||
X04145.1 | - | 7 | 739 |