Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000103.3 | NP_000094.2 | 5 | 678 | ||
NM_031226.2 | NP_112503.1 | 6 | 787 | |||
XM_005254191.1 | XP_005254248.1 | 5 | 635 | |||
GenBank mRNA | AK291778.1 | - | 6 | 785 | ||
BC020767.1 | - | 5 | 676 | |||
BC035714.1 | - | 4 | 635 | |||
BC035959.1 | - | 5 | 675 | 4770068 | ||
BC056258.1 | - | 6 | 924 | |||
BC107785.1 | - | 5 | 680 | 6663647 | ||
DQ118405.1 | - | 5 | 619 | |||
J04127.1 | - | 4 | 563 | |||
M18856.1 | - | 3 | 284 | |||
M22246.1 | - | 5 | 660 | |||
M28420.1 | - | 3 | 284 | |||
X13589.1 | - | 5 | 660 | |||
Y07508.1 | - | 4 | 585 |