Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000108.4 | NP_000099.2 | 11, 12 | 1388 | ||
NM_001289750.1 | NP_001276679.1 | 9, 10 | 1239 | |||
NM_001289751.1 | NP_001276680.1 | 10, 11 | 1319 | |||
NM_001289752.1 | NP_001276681.1 | 10, 11 | 1244 | |||
GenBank mRNA | AB209703.1 | - | 11, 12 | 1298 | ||
AK225289.1 | - | 11, 12 | 1312 | |||
AK225301.1 | - | 11, 12 | 1324 | |||
AK294146.1 | - | 12, 13 | 1325 | |||
AK295080.1 | - | 9, 10 | 1182 | |||
AK297508.1 | - | 10, 11 | 1253 | |||
AK300077.1 | - | 10, 11 | 1177 | |||
AK312346.1 | - | 11, 12 | 1335 | |||
BC018648.2 | - | 11, 12 | 1297 | 3029752 | ||
BC018696.2 | - | 11, 12 | 1297 | 3029752 | ||
J03490.1 | - | 11, 12 | 1320 | |||
J03620.1 | - | 11, 12 | 1294 |