Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000201.2 | NP_000192.2 | 7 | 2160 | ||
GenBank mRNA | AK298054.1 | - | 7 | 1767 | ||
AK298983.1 | - | 7 | 1593 | |||
AK301412.1 | - | 5 | 1242 | |||
BC015969.2 | - | 7 | 1912 | 3506766 | ||
J03132.1 | - | 7 | 1898 | |||
M24283.1 | - | 7 | 1912 | |||
M55091.1 | - | 1 | 46 |