Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000340.1 | NP_000331.1 | 11 | 2886 | ||
NM_001278658.1 | NP_001265587.1 | 10 | 2623 | |||
NM_001278659.1 | NP_001265588.1 | 10 | 2761 | |||
XM_011513087.2 | XP_011511389.1 | 12 | 2633 | |||
XM_011513089.2 | XP_011511391.1 | 8 | 2174 | |||
GenBank mRNA | BC060041.1 | - | 10 | 2506 | 4716156 | |
J03810.1 | - | 11 | 2615 |