Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000341.3 | NP_000332.2 | 10 | 2018 | ||
GenBank mRNA | AB033549.1 | - | 10 | 1999 | ||
AK091932.1 | - | 11 | 1884 | |||
AK092000.1 | - | 7 | 2124 | |||
AK223146.1 | - | 10 | 1980 | |||
AK289636.1 | - | 10 | 1996 | |||
AK313157.1 | - | 10 | 2012 | |||
BC022386.1 | - | 10 | 2016 | 4556946 | ||
BC093624.1 | - | 10 | 1977 | 7939469 | ||
BC093626.1 | - | 10 | 1972 | 7939471 | ||
DQ023512.1 | - | 7 | 2124 | |||
DQ023514.1 | - | 4 | 1141 | |||
KU178335.1 | - | 10 | 1940 | |||
L11696.1 | - | 10 | 1983 | |||
M95548.1 | - | 10 | 1984 |