Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000342.3 | NP_000333.1 | 10, 11 | 1237 | ||
XM_005257593.4 | XP_005257650.1 | 8, 9 | 1005 | |||
XM_011525129.1 | XP_011523431.1 | 10, 11 | 1264 | |||
XM_011525130.1 | XP_011523432.1 | 10, 11 | 1264 | |||
GenBank mRNA | AB526447.1 | - | 8, 9 | 1088 | ||
AM919470.1 | - | 1, 2 | 27 | |||
BC096106.3 | - | 9, 10 | 1150 | 40001948 | ||
BC096107.3 | - | 9, 10 | 1150 | 40001951 | ||
BC099628.3 | - | 9, 10 | 1150 | 40001946 | ||
BC099629.3 | - | 9, 10 | 1150 | 40001953 | ||
BC101570.1 | - | 10, 11 | 1174 | 8069076 | ||
BC101574.1 | - | 9, 10 | 1170 | 8069080 | ||
DQ419529.1 | - | 9, 10 | 1156 | |||
GQ981383.1 | - | 9, 10 | 1131 | |||
GQ981384.1 | - | 9, 10 | 1131 | |||
GQ981386.1 | - | 9, 10 | 1131 | |||
GQ981387.1 | - | 9, 10 | 1131 | |||
M27819.1 | - | 10, 11 | 1202 | |||
X12609.1 | - | 10, 11 | 1238 |