Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000343.3 | NP_000334.1 | 10 | 1332 | ||
NM_001256314.1 | NP_001243243.1 | 9 | 1050 | |||
XM_011530331.1 | XP_011528633.1 | 10 | 1314 | |||
GenBank mRNA | AK297665.1 | - | 9 | 1024 | ||
AK312948.1 | - | 10 | 1236 | |||
AK316184.1 | - | 9 | 1029 | |||
BC139839.1 | - | 10 | 1101 | |||
CR456579.1 | - | 10 | 1132 | |||
M24847.1 | - | 10 | 1092 |