Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000343.3 | NP_000334.1 | 14 | 2005 | ||
NM_001256314.1 | NP_001243243.1 | 13 | 1723 | |||
GenBank mRNA | AK297665.1 | - | 13 | 1697 | ||
AK312948.1 | - | 14 | 1909 | |||
AK316184.1 | - | 13 | 1702 | |||
BC139839.1 | - | 14 | 1774 | |||
CR456579.1 | - | 14 | 1805 | |||
M24847.1 | - | 14 | 1765 |