Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000343.3 | NP_000334.1 | 15 | 2511 | ||
NM_001256314.1 | NP_001243243.1 | 14 | 2229 | |||
GenBank mRNA | M24847.1 | - | 15 | 2270 |