Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000350.2 | NP_000341.2 | 5 | 560 | ||
GenBank mRNA | AB210040.1 | - | 5 | 546 | ||
AF000148.1 | - | 5 | 543 | |||
AF001945.1 | - | 5 | 537 | |||
AK295818.1 | - | 5 | 536 | |||
U88667.1 | - | 5 | 537 |