Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000408.4 | NP_000399.3 | 12 | 1695 | ||
NM_001083112.2 | NP_001076581.2 | 12 | 1921 | |||
XM_005246469.2 | XP_005246526.1 | 12 | 1634 | |||
XM_011510977.2 | XP_011509279.1 | 12 | 1689 | |||
XM_011510978.2 | XP_011509280.1 | 13 | 1739 | |||
XM_011510980.2 | XP_011509282.1 | 7 | 920 | |||
XM_017003830.1 | XP_016859319.1 | 12 | 1775 | |||
XM_017003831.1 | XP_016859320.1 | 11 | 1584 | |||
GenBank mRNA | AB209399.1 | - | 12 | 1690 | ||
AF311325.1 | - | 13 | 1731 | |||
AK093198.1 | - | 13 | 1730 | |||
AK290057.1 | - | 12 | 1856 | |||
AK291065.1 | - | 12 | 1934 | |||
AK292817.1 | - | 12 | 1694 | |||
AK299645.1 | - | 8 | 1036 | |||
U12424.1 | - | 12 | 1684 | |||
U36310.1 | - | 12 | 1664 |