Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000416.2 | NP_000407.1 | 4 | 582 | ||
XM_006715470.2 | XP_006715533.1 | 5 | 943 | |||
XM_006715471.2 | XP_006715534.1 | 4 | 832 | |||
XM_011535793.2 | XP_011534095.1 | 5 | 1614 | |||
XM_011535794.1 | XP_011534096.1 | 5 | 559 | |||
XM_017010827.1 | XP_016866316.1 | 5 | 653 | |||
GenBank mRNA | AF056979.1 | - | 4 | 500 | ||
AK222803.1 | - | 4 | 537 | |||
AK294252.1 | - | 5 | 625 | |||
AK297052.1 | - | 4 | 458 | |||
AK312674.1 | - | 4 | 536 | |||
BC005333.1 | - | 4 | 561 | 3950528 | ||
BT006814.1 | - | 4 | 479 | |||
EF535103.1 | - | 4 | 522 | |||
J03143.1 | - | 4 | 527 | |||
KU178144.1 | - | 4 | 479 |