Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000441.1 | NP_000432.1 | 6 | 870 | ||
XM_005250425.2 | XP_005250482.1 | 6 | 853 | |||
XM_006716025.3 | XP_006716088.1 | 5 | 649 | |||
XM_017012318.1 | XP_016867807.1 | 5 | 649 | |||
GenBank mRNA | AF030880.1 | - | 6 | 870 |