Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000441.1 | NP_000432.1 | 19 | 2377 | ||
XM_005250425.2 | XP_005250482.1 | 19 | 2360 | |||
XM_017012318.1 | XP_016867807.1 | 17 | 2078 | |||
GenBank mRNA | AF030880.1 | - | 19 | 2377 | ||
AK294388.1 | - | 10 | 1256 | |||
AK298543.1 | - | 9 | 1000 | |||
AK300604.1 | - | 10 | 1072 |