Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PECAM1 SIRNA Assays ›
Gene Symbol:
PECAM1
Gene Name:
platelet and endothelial cell adhesion molecule 1
Gene Aliases:
CD31, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, endoCAM
Chromosome Location:
Chr.17: 64319415 - 64413844 on Build GRCh38
UniGene ID:
Hs.376675
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
106105
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PECAM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000442.4
NP_000433.4
7
1456
XM_005276880.1
XP_005276937.1
7
1742
XM_005276881.1
XP_005276938.1
7
1742
XM_005276882.1
XP_005276939.1
7
1742
XM_005276883.2
XP_005276940.1
7
1742
XM_011524889.2
XP_011523191.1
8
1472
XM_011524890.1
XP_011523192.1
8
1421
XM_017024738.1
XP_016880227.1
8
1365
XM_017024739.1
XP_016880228.1
7
1742
XM_017024740.1
XP_016880229.1
8
1435
XM_017024741.1
XP_016880230.1
7
1742
GenBank mRNA
AF281287.1
-
7
1326
AF281288.1
-
7
1326
AF281289.1
-
7
1326
AF281290.1
-
7
1326
AF281291.1
-
7
1326
AF281292.1
-
7
1326
AF281293.1
-
7
1326
AF281294.1
-
7
1326
AF281295.1
-
7
1326
AF281296.1
-
7
1326
AF281297.1
-
7
1326
AF281298.1
-
7
1326
AF281299.1
-
7
1326
AF281300.1
-
7
1326
AF281301.1
-
7
1326
AF393676.1
-
7
1326
AF393677.1
-
7
1326
AF393678.1
-
7
1326
AK290692.1
-
7
1259
AK304166.1
-
6
1256
BC022512.1
-
7
1436
4816878
BC051822.1
-
7
1456
5768579
JQ287500.1
-
7
1243
M28526.1
-
7
1384
M37780.1
-
7
1368
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Gene Ontology Categories:
Function(s)
Process(es)
platelet degranulation
phagocytosis
cell adhesion
signal transduction
cell recognition
extracellular matrix organization
leukocyte migration
diapedesis
glomerular endothelium development
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline