Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000449.3 | NP_000440.1 | 11 | 2880 | ||
NM_001025603.1 | NP_001020774.1 | 11 | 2873 | |||
XM_005245405.1 | XP_005245462.1 | 11 | 2935 | |||
XM_005245406.3 | XP_005245463.1 | 11 | 2892 | |||
XM_011509847.1 | XP_011508149.1 | 11 | 2939 | |||
XM_011509848.1 | XP_011508150.1 | 11 | 2942 | |||
XM_011509849.1 | XP_011508151.1 | 11 | 2875 | |||
XM_011509850.1 | XP_011508152.1 | 11 | 3002 | |||
XM_017001999.1 | XP_016857488.1 | 11 | 3003 | |||
XM_017002000.1 | XP_016857489.1 | 11 | 3034 | |||
GenBank mRNA | AL050135.1 | - | 3 | 2115 |