Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP21A1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040090.1 | 2 | 927 | |||
GenBank mRNA | BQ637676.1 | - | 2 | 385 | ||
CD014134.1 | - | 2 | 927 |
Gene Symbol: CYP21A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000500.7 | NP_000491.4 | 4 | 604 | ||
NM_001128590.3 | NP_001122062.3 | 3 | 514 | |||
GenBank mRNA | AK054616.1 | - | 3 | 416 | ||
AK314651.1 | - | 4 | 578 | |||
AM086565.1 | - | 4 | 494 | |||
AM183945.1 | - | 4 | 494 | |||
AM183946.1 | - | 4 | 494 | |||
AM183947.1 | - | 4 | 494 | |||
AM183948.1 | - | 4 | 494 | |||
AM183949.1 | - | 4 | 494 | |||
AM183950.1 | - | 4 | 494 | |||
AM183951.1 | - | 4 | 494 | |||
BC125181.1 | - | 4 | 593 | 40122017 | ||
BC125182.1 | - | 4 | 590 | 40122018 | ||
BP198315.1 | - | 3 | 412 | |||
CD013985.1 | - | 3 | 652 | |||
CD013987.1 | - | 4 | 555 | |||
DA008890.1 | - | 4 | 503 |