Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127593.1 | NP_001121065.1 | 6 | 1424 | ||
NM_001127595.1 | NP_001121067.1 | 6 | 1406 | |||
NM_001127596.1 | NP_001121068.1 | 6 | 1403 | |||
XM_011509293.2 | XP_011507595.2 | 4 | 1055 | |||
GenBank mRNA | AK223268.1 | - | 5 | 1285 | ||
AK223295.1 | - | 5 | 1296 | |||
BC033678.1 | - | 5 | 1292 | 5180561 | ||
BC036723.1 | - | 5 | 1561 | 5206097 |