Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000578.3 | NP_000569.3 | 9, 10 | 1296 | ||
XM_005246793.3 | XP_005246850.1 | 8, 9 | 1416 | |||
XM_005246794.3 | XP_005246851.1 | 10, 11 | 1633 | |||
XM_006712709.3 | XP_006712772.1 | 9, 10 | 1490 | |||
XM_006712710.3 | XP_006712773.1 | 8, 9 | 1367 | |||
XM_006712711.3 | XP_006712774.1 | 7, 8 | 1293 | |||
XM_011511684.2 | XP_011509986.1 | 10, 11 | 1668 | |||
XM_011511685.2 | XP_011509987.1 | 9, 10 | 1525 | |||
XM_017004765.1 | XP_016860254.1 | 8, 9 | 1436 | |||
XM_017004766.1 | XP_016860255.1 | 9, 10 | 1527 | |||
GenBank mRNA | AB209650.1 | - | 7, 8 | 3010 | ||
AK298667.1 | - | 9, 10 | 1050 | |||
AK303398.1 | - | 9, 10 | 1020 | |||
AK307954.1 | - | 9, 10 | 1236 | |||
AK310914.1 | - | 9, 10 | 1052 | |||
AK311405.1 | - | 8, 9 | 903 | |||
BC033754.1 | - | 7, 8 | 1302 | |||
BC071165.1 | - | 10, 11 | 1090 | 5756382 | ||
D38171.1 | - | 7, 8 | 803 | |||
D50402.1 | - | 9, 10 | 1131 | |||
D50403.1 | - | 9, 10 | 1042 | |||
L32185.1 | - | 9, 10 | 1032 |