Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC11A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000617.2 | NP_000608.1 | 4 | 415 | ||
NM_001174125.1 | NP_001167596.1 | 4 | 407 | |||
NM_001174126.1 | NP_001167597.1 | 4 | 415 | |||
NM_001174127.1 | NP_001167598.1 | 4 | 410 | |||
NM_001174128.1 | NP_001167599.1 | 4 | 410 | |||
NM_001174129.1 | NP_001167600.1 | 4 | 675 | |||
NM_001174130.1 | NP_001167601.1 | 3 | 390 | |||
NR_033421.1 | 4 | 335 | ||||
NR_033422.1 | 4 | 410 | ||||
XM_005268911.3 | XP_005268968.1 | 4 | 467 | |||
XM_005268912.4 | XP_005268969.1 | 3 | 592 | |||
XM_011538404.2 | XP_011536706.1 | 4 | 791 | |||
XM_011538405.2 | XP_011536707.1 | 4 | 399 | |||
XM_017019355.1 | XP_016874844.1 | 4 | 675 | |||
XM_017019356.1 | XP_016874845.1 | 4 | 605 | |||
GenBank mRNA | AB004857.1 | - | 4 | 358 | ||
AB062284.1 | - | 4 | 332 | |||
AF046997.1 | - | 4 | 368 | |||
AF064484.1 | - | 4 | 358 | |||
AJ493662.1 | - | 4 | 407 | |||
AK094735.1 | - | 4 | 675 | |||
AK128640.1 | - | 4 | 389 | |||
AK296445.1 | - | 3 | 369 | |||
AK315987.1 | - | 4 | 321 | |||
AK316507.1 | - | 3 | 346 | |||
BC002592.2 | - | 4 | 362 | 3160522 | ||
BC100014.1 | - | 4 | 383 | 5300266 | ||
DA417090.1 | - | 4 | 415 | |||
DA737206.1 | - | 4 | 673 | |||
DA957633.1 | - | 4 | 390 | |||
DC324894.1 | - | 3 | 390 | |||
KT455015.1 | - | 4 | 371 | |||
L37347.1 | - | 1 | 108 |