Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEACAM4 SIRNA Assays ›
Gene Symbol:
CEACAM4
Gene Name:
carcinoembryonic antigen related cell adhesion molecule 4
Gene Aliases:
CGM7, CGM7_HUMAN, NCA
Chromosome Location:
Chr.19: 41616986 - 41627087 on Build GRCh38
UniGene ID:
Hs.12
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
10618
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEACAM4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001817.3
NP_001808.2
2
471
XM_006722977.3
XP_006723040.1
2
484
XM_011526343.1
XP_011524645.1
2
471
XM_011526345.2
XP_011524647.1
2
484
XM_011526348.2
XP_011524650.1
2
471
XM_011526349.2
XP_011524651.1
2
484
XM_011526350.2
XP_011524652.1
2
471
XM_011526352.1
XP_011524654.1
2
471
XM_011526354.1
XP_011524656.1
2
471
XM_017026199.1
XP_016881688.1
2
471
XM_017026200.1
XP_016881689.1
2
471
XM_017026201.1
XP_016881690.1
2
484
XM_017026202.1
XP_016881691.1
2
484
XM_017026203.1
XP_016881692.1
2
484
XM_017026204.1
XP_016881693.1
2
484
XM_017026205.1
XP_016881694.1
2
471
XM_017026206.1
XP_016881695.1
2
471
XM_017026207.1
XP_016881696.1
2
484
XM_017026208.1
XP_016881697.1
2
484
XM_017026209.1
XP_016881698.1
2
471
XM_017026210.1
XP_016881699.1
2
471
XM_017026211.1
XP_016881700.1
2
471
XM_017026212.1
XP_016881701.1
2
471
GenBank mRNA
AB924369.1
-
2
360
AB924370.1
-
2
360
AK298701.1
-
2
471
BX644557.1
-
2
472
D90276.1
-
2
457
Back To Top
More Information
Gene Ontology Categories:
Function(s)
phagocytosis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline