Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001821.3 | NP_001812.2 | 1 | 305 | ||
GenBank mRNA | AK314125.1 | - | 1 | 569 | ||
BC117360.1 | - | 1 | 175 | 40125911 | ||
BM470658.1 | - | 101 | ||||
X64728.1 | - | 1 | 305 |