Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000745.3 | NP_000736.2 | 5 | 641 | ||
NM_001307945.1 | NP_001294874.1 | 5 | 641 | |||
XM_005254142.3 | XP_005254199.1 | 5 | 640 | |||
XM_017021881.1 | XP_016877370.1 | 5 | 640 | |||
GenBank mRNA | AF385586.1 | - | 5 | 587 | ||
BC033639.1 | - | 5 | 603 | 5501542 | ||
BM839817.1 | - | 5 | 642 | |||
BX332469.2 | - | 5 | 588 | |||
M83712.1 | - | 5 | 589 | |||
U62434.1 | - | 5 | 595 | |||
Y08419.1 | - | 5 | 441 |