Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000745.3 | NP_000736.2 | 5 | 890 | ||
XM_005254142.3 | XP_005254199.1 | 5 | 889 | |||
XM_017021881.1 | XP_016877370.1 | 5 | 889 | |||
GenBank mRNA | AF385586.1 | - | 5 | 836 | ||
BC033639.1 | - | 5 | 852 | 5501542 | ||
M83712.1 | - | 5 | 838 | |||
U62434.1 | - | 5 | 844 | |||
Y08419.1 | - | 5 | 690 |