Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000781.2 | NP_000772.2 | 6, 7 | 1309 | ||
NM_001099773.1 | NP_001093243.1 | 6, 7 | 1383 | |||
GenBank mRNA | AK056794.1 | - | 6, 7 | 1378 | ||
AK225715.1 | - | 5, 6 | 897 | |||
AK292300.1 | - | 6, 7 | 1215 | |||
AK292301.1 | - | 5, 6 | 1173 | |||
AK293211.1 | - | 7, 8 | 1175 | |||
AK300178.1 | - | 6, 7 | 1125 | |||
AY603498.1 | - | 2, 3 | 197 | |||
BC032329.1 | - | 6, 7 | 1198 | 5165263 | ||
KU178011.1 | - | 6, 7 | 1154 | |||
M14565.1 | - | 6, 7 | 1198 | |||
M28253.1 | - | 2, 3 | 308 |