Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP24A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000782.4 | NP_000773.2 | 9, 10 | 1640 | ||
XM_005260304.4 | XP_005260361.1 | 9, 10 | 1648 | |||
XM_017027691.1 | XP_016883180.1 | 9, 10 | 1648 | |||
XM_017027692.1 | XP_016883181.1 | 9, 10 | 1648 | |||
GenBank mRNA | AY858838.1 | - | 7, 8 | 1076 | ||
L13286.1 | - | 9, 10 | 1632 | |||
S67623.1 | - | 4, 5 | 473 |