Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000812.3 | NP_000803.2 | 9 | 1808 | ||
XM_017007985.1 | XP_016863474.1 | 5 | 946 | |||
GenBank mRNA | AK225167.1 | - | 9 | 1485 | ||
AK225686.1 | - | 10 | 1455 | |||
AK293691.1 | - | 8 | 1385 | |||
BC022449.1 | - | 9 | 1512 | 4797401 | ||
X14767.1 | - | 9 | 1465 |