Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000829.3 | NP_000820.3 | 8 | 1477 | ||
NM_001077243.2 | NP_001070711.2 | 8 | 1477 | |||
NM_001077244.1 | NP_001070712.1 | 8 | 1477 | |||
NM_001112812.1 | NP_001106283.1 | 7 | 1345 | |||
NR_046356.1 | 7 | 1345 | ||||
XM_005271518.3 | XP_005271575.1 | 8 | 1521 | |||
XM_006718823.1 | XP_006718886.1 | 8 | 1151 | |||
XM_011542775.2 | XP_011541077.1 | 8 | 1521 | |||
XM_011542776.2 | XP_011541078.1 | 6 | 3055 | |||
XM_011542777.2 | XP_011541079.1 | 5 | 2925 | |||
XM_017017609.1 | XP_016873098.1 | 8 | 1521 | |||
XM_017017610.1 | XP_016873099.1 | 8 | 1521 | |||
XM_017017611.1 | XP_016873100.1 | 5 | 1162 | |||
GenBank mRNA | AB209092.1 | - | 7 | 1315 | ||
BC045546.1 | - | 8 | 1471 | |||
BC114549.1 | - | 7 | 1030 | |||
BC142654.1 | - | 8 | 1425 | |||
BC146816.1 | - | 7 | 1304 | |||
BC150209.1 | - | 8 | 1425 | |||
U16129.1 | - | 7 | 1031 |