Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000829.3 | NP_000820.3 | 9 | 1529 | ||
NM_001077243.2 | NP_001070711.2 | 9 | 1529 | |||
NM_001077244.1 | NP_001070712.1 | 9 | 1529 | |||
NM_001112812.1 | NP_001106283.1 | 8 | 1397 | |||
NR_046356.1 | 8 | 1397 | ||||
XM_005271518.3 | XP_005271575.1 | 9 | 1573 | |||
XM_006718823.1 | XP_006718886.1 | 9 | 1203 | |||
XM_011542775.2 | XP_011541077.1 | 9 | 1573 | |||
XM_011542776.2 | XP_011541078.1 | 7 | 3107 | |||
XM_011542777.2 | XP_011541079.1 | 6 | 2977 | |||
XM_017017609.1 | XP_016873098.1 | 9 | 1573 | |||
XM_017017610.1 | XP_016873099.1 | 9 | 1573 | |||
XM_017017611.1 | XP_016873100.1 | 6 | 1214 | |||
GenBank mRNA | AB209092.1 | - | 8 | 1367 | ||
BC045546.1 | - | 9 | 1523 | |||
BC114549.1 | - | 8 | 1082 | |||
BC142654.1 | - | 9 | 1477 | |||
BC146816.1 | - | 8 | 1356 | |||
BC150209.1 | - | 9 | 1477 | |||
U16129.1 | - | 8 | 1083 |