Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000834.3 | NP_000825.2 | 6 | 1531 | ||
XM_011520628.2 | XP_011518930.1 | 7 | 2324 | |||
XM_011520629.2 | XP_011518931.1 | 8 | 2131 | |||
XM_017019219.1 | XP_016874708.1 | 7 | 1879 | |||
GenBank mRNA | AB208850.1 | - | 3 | 310 | ||
BC113618.1 | - | 6 | 1430 | 8322670 | ||
BC113620.1 | - | 6 | 1430 | 8322672 | ||
U11287.1 | - | 6 | 1562 | |||
U88963.1 | - | 6 | 1531 | |||
U90278.1 | - | 6 | 1561 |