Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000834.3 | NP_000825.2 | 13 | 5702 | ||
XM_005253351.3 | XP_005253408.1 | 4 | 3429 | |||
XM_011520628.2 | XP_011518930.1 | 14 | 6495 | |||
XM_011520629.2 | XP_011518931.1 | 15 | 6302 | |||
XM_017019219.1 | XP_016874708.1 | 14 | 6050 | |||
GenBank mRNA | U11287.1 | - | 13 | 5731 | ||
U88963.1 | - | 13 | 5700 |