Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000878.3 | NP_000869.1 | 10 | 1276 | ||
GenBank mRNA | AK312860.1 | - | 10 | 1270 | ||
BC025691.1 | - | 10 | 1259 | 5207833 | ||
CR456506.1 | - | 9 | 1164 | |||
M26062.1 | - | 10 | 1276 |