Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000903.2 | NP_000894.1 | 6 | 2351 | ||
NM_001025433.1 | NP_001020604.1 | 5 | 2249 | |||
NM_001025434.1 | NP_001020605.1 | 5 | 2237 | |||
NM_001286137.1 | NP_001273066.1 | 4 | 2173 | |||
GenBank mRNA | AV729122.1 | - | 1 | 106 | ||
BM828301.1 | - | 1 | 580 | |||
J03934.1 | - | 6 | 2210 |