Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAM SIRNA Assays ›
Gene Symbol:
PAM
Gene Name:
peptidylglycine alpha-amidating monooxygenase
Gene Aliases:
PAL, PHM
Chromosome Location:
Chr.5: 102754783 - 103031105 on Build GRCh38
UniGene ID:
Hs.369430
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
106325
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000919.3
NP_000910.2
18
2215
NM_001177306.1
NP_001170777.1
18
2215
NM_001319943.1
NP_001306872.1
20
2835
NM_138766.2
NP_620121.1
18
2215
NM_138821.2
NP_620176.1
17
1894
NM_138822.2
NP_620177.1
18
2215
NR_033440.1
17
2157
XM_011543419.2
XP_011541721.1
20
2389
XM_017009495.1
XP_016864984.1
19
2265
XM_017009496.1
XP_016864985.1
19
2265
XM_017009497.1
XP_016864986.1
19
2335
XM_017009498.1
XP_016864987.1
19
2265
XM_017009499.1
XP_016864988.1
18
2211
XM_017009500.1
XP_016864989.1
18
2211
XM_017009501.1
XP_016864990.1
18
2211
XM_017009502.1
XP_016864991.1
19
2265
XM_017009503.1
XP_016864992.1
19
2265
XM_017009504.1
XP_016864993.1
18
2312
XM_017009505.1
XP_016864994.1
19
2335
XM_017009506.1
XP_016864995.1
19
2265
XM_017009507.1
XP_016864996.1
18
2014
XM_017009508.1
XP_016864997.1
17
1890
XM_017009509.1
XP_016864998.1
17
1890
XM_017009510.1
XP_016864999.1
17
1994
XM_017009511.1
XP_016865000.1
19
2265
XM_017009512.1
XP_016865001.1
19
2265
XM_017009513.1
XP_016865002.1
19
2265
XM_017009514.1
XP_016865003.1
17
1890
XM_017009515.1
XP_016865004.1
17
1890
XM_017009516.1
XP_016865005.1
17
1890
GenBank mRNA
AB095007.1
-
18
1842
AF010472.1
-
17
1758
AK290158.1
-
18
1953
AK296521.1
-
17
1923
BC018127.1
-
17
1894
3887394
BT007419.1
-
17
1521
M37721.1
-
18
2030
S75037.1
-
18
1922
S75038.1
-
18
1922
Back To Top
More Information
Panther Classification:
Molecular Function -
oxygenase
Gene Ontology Categories:
Function(s)
Process(es)
peptide amidation
response to hypoxia
long-chain fatty acid metabolic process
regulation of transcription from RNA polymerase II promoter
central nervous system development
heart development
lactation
response to pH
toxin metabolic process
protein amidation
ovulation cycle process
response to estradiol
regulation of actin cytoskeleton organization
odontogenesis
response to drug
response to copper ion
regulation of protein secretion
protein homooligomerization
response to glucocorticoid
oxidation-reduction process
maternal process involved in female pregnancy
limb development
peptidylglycine monooxygenase activity
peptidylamidoglycolate lyase activity
copper ion binding
calcium ion binding
protein binding
zinc ion binding
protein kinase binding
L-ascorbic acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline