Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000940.2 | NP_000931.1 | 8 | 877 | ||
GenBank mRNA | AF320003.1 | - | 8 | 852 | ||
AF329433.1 | - | 8 | 852 | |||
AK096114.1 | - | 3 | 2208 | |||
BC070374.1 | - | 9 | 863 | 4710338 | ||
CD014095.1 | - | 9 | 1009 | |||
CD014097.1 | - | 8 | 813 | |||
CD014100.1 | - | 8 | 736 | |||
L48516.1 | - | 8 | 813 |