Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000949.6 | NP_000940.1 | 10 | 1444 | ||
NM_001204314.2 | NP_001191243.1 | 7 | 819 | |||
XM_006714484.2 | XP_006714547.1 | 10 | 1262 | |||
XM_011514068.2 | XP_011512370.1 | 10 | 1277 | |||
XM_011514069.2 | XP_011512371.1 | 10 | 1216 | |||
XM_017009645.1 | XP_016865134.1 | 9 | 1210 | |||
GenBank mRNA | AF349939.1 | - | 6 | 714 | ||
AK313270.1 | - | 10 | 1332 | |||
BC059392.1 | - | 10 | 1330 | 30343995 | ||
M31661.1 | - | 10 | 1301 |